Merck
  • Molecular action of the type 1 ribosome-inactivating protein saporin 5 on Vicia sativa ribosomes.
Molecular action of the type 1 ribosome-inactivating protein saporin 5 on Vicia sativa ribosomes.
FEBS letters (1993-07-05)
R Iglesias, F J Arias, M A Rojo, C Escarmis, J M Ferreras, T Girbés
ABSTRACT
The type 1 ribosome-inactivating protein (RIP) saporin 5 isolated from seeds of Saponaria officinalis L. strongly inhibited translation carried out by Vicia sativa L. purified ribosomes. The toxin multidepurinated V. sativa rRNA, which upon treatment with acid aniline releases several RNA fragments including an RNA fragment of approximately 370 nucleotides the 5'-end sequence of which was 5'-GAGGAACG-3'.
Sigma-Aldrich
Saporin from Saponaria officinalis seeds, lyophilized powder
Pricing and availability is not currently available.