Merck
Tetrahydrobenzotriazines as a new class of nematocide.
Bioorganic & medicinal chemistry (2006-11-28)
Keiji Nishiwaki, Azusa Okamoto, Keizo Matsuo, Yoshio Hayase, Shunichiro Masaki, Riichi Hasegawa, Katsuaki Ohba
ABSTRACT
Tetrahydrobenzotriazines, a novel class of heterocyclic compounds, were synthesized. Examination of their biological activities resulted in the discovery that some of them possess potent nematocidal activity.
Product Description
Supelco
Methyl isothiocyanate, PESTANAL®, analytical standard
Sigma-Aldrich
Methyl isothiocyanate, 97%