Merck
  • Simultaneous quantitation of hydroxychloroquine and its metabolites in mouse blood and tissues using LC-ESI-MS/MS: An application for pharmacokinetic studies.
Simultaneous quantitation of hydroxychloroquine and its metabolites in mouse blood and tissues using LC-ESI-MS/MS: An application for pharmacokinetic studies.
Journal of chromatography. B, Analytical technologies in the biomedical and life sciences (2017-12-06)
Yashpal S Chhonker, Richard L Sleightholm, Jing Li, David Oupický, Daryl J Murry
ABSTRACT
Hydroxychloroquine (HCQ) has been shown to disrupt autophagy and sensitize cancer cells to radiation and chemotherapeutic agents. However, the optimal delivery method, dose, and tumor concentrations required for these effects are not known. This is in part due to a lack of sensitive and reproducible analytical methods for HCQ quantitation in small animals. As such, we developed and validated a selective and sensitive liquid chromatography coupled with tandem mass spectrometry (LC-MS/MS) method for simultaneous quantitation of hydroxychloroquine and its metabolites in mouse blood and tissues. The chromatographic separation and detection of analytes were achieved on a reversed phase Thermo Aquasil C18 (50×4.6mm, 3μ) column, with gradient elution using 0.2% formic acid and 0.1% formic acid in methanol as mobile phase at a flow rate of 0.5mL/min. Simple protein precipitation was utilized for extraction of analytes from the desired matrix. Analytes were separated and quantitated using MS/MS with an electrospray ionization source in positive multiple reaction monitoring (MRM) mode. The MS/MS response was linear over the concentration range from 1 to 2000ng/mL for all analytes with a correlation coefficient (R2) of 0.998 or better. The within- and between-day precision (relative standard deviation, % RSD) and accuracy were within the acceptable limits per FDA guidelines. The validated method was successfully applied to a preclinical pharmacokinetic mouse study involving low volume blood and tissue samples for hydroxychloroquine and metabolites.