Merck
  • Endoscopic transpapillary gallbladder drainage for the management of acute calculus cholecystitis patients unfit for urgent cholecystectomy.
Endoscopic transpapillary gallbladder drainage for the management of acute calculus cholecystitis patients unfit for urgent cholecystectomy.
PloS one (2020-10-10)
Tae Hyeon Kim, Dong Eun Park, Hyung Ku Chon
ABSTRACT
Endoscopic transpapillary gallbladder drainage (ETGBD) has been proposed as an alternative to surgery or percutaneous cholecystostomy in patients with acute calculus cholecystitis (ACC). We aimed to evaluate the safety and efficacy of ETGBD via endoscopic transpapillary gallbladder stenting (ETGBS) or endoscopic naso-gallbladder drainage (ENGBD) as either a bridging or a definitive treatment option for patients with ACC when a cholecystectomy is delayed or cannot be performed. From July 2014 to December 2018, 171 patients with ACC in whom ETGBD were attempted were retrospectively reviewed. The technical and clinical success rates and adverse events were evaluated. Moreover, the predictive factors for technical success and the stent patency in the ETGBS group with high surgical risk were examined. The technical and clinical success rates by intention-to-treat analysis for ETGBD were 90.6% (155/171) and 90.1% (154/171), respectively. Visible cystic duct on cholangiography were significant technical success predictor (adjusted odds ratio: 7.099, 95% confidence interval: 1.983-25.407, P = 0.003) as per logistic regression analysis. Adverse events occurred in 12.2% of patients (21/171: mild pancreatitis, n = 9; acute cholangitis, n = 6; post-endoscopic sphincterotomy bleeding, n = 4; and stent migration, n = 1; ACC recurrence, n = 1), but all patients were treated with conservative management and endoscopic treatment. Among the ETGBS group, the median stent patency in 70 patients with high surgical risk was 503 days (interquartile range: 404.25-775 days). ETGBD, using either ETGBS or ENGBD, may be a suitable bridging option for ACC patients unfit for urgent cholecystectomy. In high surgical risk patients, ETGBS may be a promising and useful treatment modality with low ACC recurrence.
Sigma-Aldrich
1,1,3,3-Tetramethyldisilazane, 97%