Merck
  • Differential influences of the BPA, BPS and BPF on in vitro IL-17 secretion by mouse and human T cells.
Differential influences of the BPA, BPS and BPF on in vitro IL-17 secretion by mouse and human T cells.
Toxicology in vitro : an international journal published in association with BIBRA (2020-09-11)
Yann Malaisé, Hélène Le Mentec, Lydie Sparfel, Laurence Guzylack-Piriou
ABSTRACT
The endocrine disruptor and food contaminant bisphenol A (BPA) is frequently present in consumer plastics and can produce several adverse health effects participating in the development of inflammatory and autoimmune diseases. Regulatory restrictions have been established to prevent risks for human health, leading to the substitution of BPA by structural analogues, such as bisphenol S (BPS) and F (BPF). In this study, we aimed at comparing the in vitro impact of these bisphenols from 0.05 to 50,000 nM on Th17 differentiation, frequency and function in mouse systemic and intestinal immune T cells and in human blood T cells. This study reports the ability of these bisphenols, at low and environmentally relevant concentration, i.e, 0.05 nM, to increase significantly IL-17 production in mouse T cells but not in human T lymphocytes. The use of an aryl hydrocarbon receptor (AhR) specific inhibitor demonstrated its involvement in this bisphenol-induced IL-17 production. We also observed an increased IL-17 secretion by BPS and BPF, and not by BPA, in mouse naive T cells undergoing in vitro Th17 differentiation. In total, this study emphasizes the link between bisphenol exposures and the susceptibility to develop immune diseases, questioning thus the rational of their use to replace BPA.
Sigma-Aldrich
FICZ, ≥95% (HPLC)