Merck
  • Intramacrophage ROS Primes the Innate Immune System via JAK/STAT and Toll Activation.
Intramacrophage ROS Primes the Innate Immune System via JAK/STAT and Toll Activation.
Cell reports (2020-11-12)
Sveta Chakrabarti, Sandhya S Visweswariah
ABSTRACT
Tissue injury is one of the most severe environmental perturbations for a living organism. When damage occurs in adult Drosophila, there is a local response of the injured tissue and a coordinated action across different tissues to help the organism overcome the deleterious effect of an injury. We show a change in the transcriptome of hemocytes at the site of tissue injury, with pronounced activation of the Toll signaling pathway. We find that induction of the cytokine upd-3 and Toll receptor activation occur in response to injury alone, in the absence of a pathogen. Intracellular accumulation of hydrogen peroxide in hemocytes is essential for upd-3 induction and is facilitated by the diffusion of hydrogen peroxide through a channel protein Prip. Importantly, hemocyte activation and production of reactive oxygen species (ROS) at the site of a sterile injury provide protection to flies on subsequent infection, demonstrating training of the innate immune system.
Sigma-Aldrich
Dulbecco′s Phosphate Buffered Saline, Modified, without calcium chloride and magnesium chloride, powder, suitable for cell culture
Sigma-Aldrich
Bismarck Brown R, for microscopy (Bact., Hist.)
Sigma-Aldrich
Propyl gallate, powder
Sigma-Aldrich
DAPI, for nucleic acid staining