Merck
  • Fat mass and obesity-associated protein promotes the tumorigenesis and development of liver cancer.
Fat mass and obesity-associated protein promotes the tumorigenesis and development of liver cancer.
Oncology letters (2020-07-30)
Ziqi Ye, Shibing Wang, Wanyuan Chen, Xin Zhang, Jie Chen, Jinying Jiang, Mingshan Wang, Li Zhang, Zixue Xuan
ABSTRACT
Liver cancer is the fourth leading cause of cancer-associated mortality worldwide. Statistics indicate that the incidence of liver cancer has been increasing and that its prognosis remains poor. Fat mass and obesity-associated protein (FTO) is a demethylase that is involved in N6-methyladenosine (m6a) RNA modification; however, to the best of our knowledge, its role in tumorigenesis and development of liver cancer remains unknown. In the present study, cell proliferation, colony formation, apoptosis, Transwell and wound healing assays of small interfering (si)RNA-FTO HepG2 cells were performed, and the levels of m6A RNA methylation were assessed. Additionally, the prognostic value of FTO in liver cancer was analyzed using immunohistochemistry analysis. The results from the EpiQuik m6A RNA methylation quantitative assay revealed that knockdown of FTO increased the total m6A methylation level. Notably, FTO promoted the proliferation and migration of liver cancer cells. Additionally, FTO expression was upregulated in patients with liver cancer and was associated with a high Edmondson Grade, which served as an independent prognostic factor for liver cancer. Results from the Kaplan-Meier survival analysis revealed that low expression levels of FTO predicted a good prognosis. The 5-year overall survival of the low FTO expression group was 68% compared with 48% in the high FTO expression group (P=0.077). In conclusion, the present study suggested that FTO regulates the tumorigenesis and development of liver cancer.