Merck
  • Mechanistic Aspects of Condensed- and Gaseous-Phase Activities of Some Phosphorus-Containing Fire Retardants.
Mechanistic Aspects of Condensed- and Gaseous-Phase Activities of Some Phosphorus-Containing Fire Retardants.
Polymers (2020-08-17)
Ananya Thomas, Malavika Arun, Khalid Moinuddin, Paul Joseph
ABSTRACT
As a part of our ongoing investigations on passively fire protecting polymeric materials, we have been employing both reactive and additive routes involving phosphorus-containing compounds. These included inorganic and organic substances, and in the latter case, the phosphorus-bearing groups differed in terms of the chemical environments (phosphite, phosphate, phosphine, phosphine oxide and phosphonate ester) and oxidation state of the P atom (i.e., III, or V). The overall flammability profiles of wood substrates coated with the phosphorus-containing compounds were obtained through cone calorimetric measurements. The elemental composition, morphology and chemical natures of the char residues, obtained from the cone tests, were analysed through a variety of spectroscopic, chromatographic and spectrometric means. From the complementary information, obtained through these analyses, some probable mechanistic pathways that underpin the condensed- and gaseous-phase activities of the different additives are suggested. It was found that the inorganic solid additive, i.e., (NH4)2HPO4, underwent a two-step degradation, yielding ammonia gas and phosphoric acid. Furthermore, the liquid additives, owing to their volatility as compared to the solid ones, showed a relatively higher presence in the vapour phase than volatile fragments emanating from the latter ones (i.e., from phosphine and the phosphine oxides).
Sigma-Aldrich
Diethyl benzylphosphonate, 99%