Merck
  • Preparation of efficient photothermal materials from waste coffee grounds for solar evaporation and water purification.
Preparation of efficient photothermal materials from waste coffee grounds for solar evaporation and water purification.
Scientific reports (2020-07-31)
Chih-Feng Wang, Chih-Lin Wu, Shiao-Wei Kuo, Wei-Song Hung, Kuo-Jung Lee, Hsieh-Chih Tsai, Chi-Jung Chang, Juin-Yih Lai
ABSTRACT
Effective water use is currently a critical global challenge needed to prevent water shortages and has attracted significant research attention. The realization of solar-driven water evaporation by using effective converters has attracted considerable attention in recent years owing to its potential for seawater desalination and wastewater treatment. Consequently, this paper proposes a simple two-step method to prepare low-cost and self-floating photothermal converters from waste coffee grounds. First, the coffee grounds were carbonized at 1,000 °C to develop broadband absorption, and the carbonized coffee grounds were modified using hydrophobic silane to enhance the water-floatation property of the grounds. The prepared hydrophobic carbonized coffee grounds exhibited good performance for desalination and water purification under sunlight irradiation. The self-floatation ability, low cost, well solar evaporation performance, and easy preparation contribute to the promising potential of using hydrophobic carbonized coffee grounds infuture steps toward eco-friendly desalination procedures.
Sigma-Aldrich
Dodecyltriethoxysilane, technical
Pricing and availability is not currently available.