Merck
  • Binary nonmetal S and P-co-doping into mesoporous PtPd nanocages boosts oxygen reduction electrocatalysis.
Binary nonmetal S and P-co-doping into mesoporous PtPd nanocages boosts oxygen reduction electrocatalysis.
Nanoscale (2020-07-08)
Shuli Yin, You Xu, Songliang Liu, Hongjie Yu, Ziqiang Wang, Xiaonian Li, Liang Wang, Hongjing Wang
ABSTRACT
The development of doped noble metal catalysts with nonmetal elements to improve the catalytic performance toward the oxygen reduction reaction (ORR) is significant for proton exchange membrane fuel cell technology. Here, we report a one-pot for dual-nonmetal-doping strategy for the synthesis of S and P-co-doped mesoporous PtPd nanocages (PtPdSP mNCs) by using pre-synthesized mesoporous PtPd nanocages (PtPd mNCs) as the precursor and triphenylphosphine sulphide as both S and P sources. Benefitting from the combined advantages of metal-nonmetal incorporation, hollow cavity and surface porosity, the resultant quaternary PtPdSP mNCs exhibit outstanding ORR activity and long-term stability. This research work provides a good strategy for the doping of two or more selected nonmetallic elements into metallic nanocrystals with a controllable structure and composition.
Sigma-Aldrich
Isopropyl alcohol, natural, ≥98%, FG
Sigma-Aldrich
Triphenylphosphine sulfide, 98%