Merck
  • Diazonium salt-derived 4-(dimethylamino)phenyl groups as hydrogen donors in surface-confined radical photopolymerization for bioactive poly(2-hydroxyethyl methacrylate) grafts.
Diazonium salt-derived 4-(dimethylamino)phenyl groups as hydrogen donors in surface-confined radical photopolymerization for bioactive poly(2-hydroxyethyl methacrylate) grafts.
Langmuir : the ACS journal of surfaces and colloids (2012-05-03)
Sarra Gam-Derouich, Aazdine Lamouri, Catherine Redeuilh, Philippe Decorse, François Maurel, Benjamin Carbonnier, Selim Beyazıt, Gorkem Yilmaz, Yusuf Yagci, Mohamed M Chehimi
ABSTRACT
In this paper we describe a novel methodology for grafting polymers via radical photopolymerization initiated on gold surfaces by aryl layers from diazonium salt precursors. The parent 4-(dimethylamino)benzenediazonium salt was electroreduced on a gold surface to provide 4-(dimethylamino)phenyl (DMA) hydrogen donor layers; free benzophenone in solution was used as a photosensitizer to strip hydrogen from the grafted DMA. This system permitted efficient surface initiation of photopolymerization of 2-hydroxyethyl methacrylate. The resulting poly(2-hydroxyethyl methacrylate) (PHEMA) grafts were found to be very adherent to the surface as they resist total failure after being soaked in the well-known paint stripper methyl ethyl ketone. The PHEMA grafts were reacted with 1,1'-carbonyldiimidazole to yield carbamate groups that are able to react readily with amino groups from proteins. The final surface consisted of protein-functionalized PHEMA grafts where bovine serum albumin (BSA) protein is specifically linked to the grafts by covalent bonds. We used X-ray photoelectron spectroscopy to monitor the chemical changes at the gold surface all along the process from the neat gold to the end-protein-functionalized polymer grafts: the PHEMA graft thickness ranged from 7 to 27 nm, and the activation by 1,1'-carbonyldiimidazole reached 37% of the OH groups, which was sufficient for 90% surface coverage of the grafts by BSA. This work conclusively provides a new approach for bridging reactive and functional polymers to surfaces via aryl diazonium salts in a simple, fast, and efficient approach of importance in biomedical and other applications.
Sigma-Aldrich
2-Hydroxyethyl methacrylate, embedding medium (for microscopy)