Merck
  • Indirect mechanism of oestradiol stimulation of cell proliferation of human breast cancer cell lines.
Indirect mechanism of oestradiol stimulation of cell proliferation of human breast cancer cell lines.
British journal of cancer (1986-01-01)
A E Lykkesfeldt, P Briand
ABSTRACT
The human breast cancer cell line MCF-7 requires oestrogen to produce and promote growth of tumours in athymic mice. In vitro, however, MCF-7 cells proliferate rapidly without supply of oestrogen (Briand & Lykkesfeldt, 1984). Oestrogen stimulation of proliferation of MCF-7 cells can be achieved when the cells are grown at high concentration of newborn calf serum (NCS, 10%) or oestrogen deprived foetal calf serum (10%). The stimulation involves an abolishment of inhibitory activity present in the serum. The oestradiol stimulated cultures grow rapidly for a much longer time period and attain a much higher cell density than the unstimulated cultures. Oestrogen is specific for the promotion of cell proliferation and only oestrogen receptor positive cell lines with a functional oestrogen receptor mechanism can be stimulated. We assume that oestradiol acts directly on the cells and via the oestrogen receptor mechanism induces the synthesis of a substance which abolishes the inhibitory activity in serum. We call this mechanism of action an indirect stimulation of cell proliferation. A similar mechanism may exist in vivo since we find that serum from athymic mice contains a growth inhibitory activity towards MCF-7 cells and the inhibitory effect can be abolished by oestradiol.
Sigma-Aldrich
MCF-7/S0.5 Human Breast Cancer Cell Line, MCF-7/S0.5 is a tamoxifen-sensitive cell line established from the MCF-7 human breast cancer cell line by adaption to growth in low serum conditions.