Merck
  • Structural and Functional Analysis of a β2-Adrenergic Receptor Complex with GRK5.
Structural and Functional Analysis of a β2-Adrenergic Receptor Complex with GRK5.
Cell (2017-04-22)
Konstantin E Komolov, Yang Du, Nguyen Minh Duc, Robin M Betz, João P G L M Rodrigues, Ryan D Leib, Dhabaleswar Patra, Georgios Skiniotis, Christopher M Adams, Ron O Dror, Ka Young Chung, Brian K Kobilka, Jeffrey L Benovic
ABSTRACT
The phosphorylation of agonist-occupied G-protein-coupled receptors (GPCRs) by GPCR kinases (GRKs) functions to turn off G-protein signaling and turn on arrestin-mediated signaling. While a structural understanding of GPCR/G-protein and GPCR/arrestin complexes has emerged in recent years, the molecular architecture of a GPCR/GRK complex remains poorly defined. We used a comprehensive integrated approach of cross-linking, hydrogen-deuterium exchange mass spectrometry (MS), electron microscopy, mutagenesis, molecular dynamics simulations, and computational docking to analyze GRK5 interaction with the β2-adrenergic receptor (β2AR). These studies revealed a dynamic mechanism of complex formation that involves large conformational changes in the GRK5 RH/catalytic domain interface upon receptor binding. These changes facilitate contacts between intracellular loops 2 and 3 and the C terminus of the β2AR with the GRK5 RH bundle subdomain, membrane-binding surface, and kinase catalytic cleft, respectively. These studies significantly contribute to our understanding of the mechanism by which GRKs regulate the function of activated GPCRs. PAPERCLIP.