Merck
  • Alpha-hydroxylation of lignoceroyl-CoA in rat brain microsomes: involvement of NADPH-cytochrome c reductase and topical distribution.
Alpha-hydroxylation of lignoceroyl-CoA in rat brain microsomes: involvement of NADPH-cytochrome c reductase and topical distribution.
The International journal of biochemistry (1990-01-01)
H Shigematsu, Y Hisanari, Y Kishimoto
ABSTRACT
1. The enzymatic mechanism of the alpha-hydroxylation of lignoceroyl-CoA, an intermediate in the synthesis of hydroxyceramide, was studied. In the presence of NADPH, sphingosine and microsomes from 20-day-old rat brain, 14C from [1-14C]lignoceroyl-CoA was incorporated into hydroxyceramide. 2. The alpha-hydroxylation of lignoceroyl-CoA in rat brain microsomes was strongly inhibited by a rabbit anti-immunoglobulin G which was prepared against rat liver microsomal NADPH-cytochrome c reductase. However, anti-immunoglobulin G against cytochrome b5 did not inhibit the alpha-hydroxylase activity. 3. The alpha-hydroxylation activity was more sensitive to trypsin treatment than was NADPH-cytochrome c reductase in rat brain microsomes. This indicates that either alpha-hydroxylase itself or an unknown factor essential in alpha-hydroxylation is highly exposed to the surface of brain microsomes.