Merck
  • Characterization of a small molecule that promotes cell cycle activation of human induced pluripotent stem cell-derived cardiomyocytes.
Characterization of a small molecule that promotes cell cycle activation of human induced pluripotent stem cell-derived cardiomyocytes.
Journal of molecular and cellular cardiology (2019-01-27)
Masamichi Ito, Hironori Hara, Norifumi Takeda, Atsuhiko T Naito, Seitaro Nomura, Masaki Kondo, Yutaka Hata, Masanobu Uchiyama, Hiroyuki Morita, Issei Komuro
ABSTRACT
Since regenerative capacity of adult mammalian myocardium is limited, activation of the endogenous proliferative capacity of existing cardiomyocytes is a potential therapeutic strategy for treating heart diseases accompanied by cardiomyocyte loss. Recently, we performed a compound screening and developed a new drug named TT-10 (C11H10FN3OS2) which promotes the proliferation of murine cardiomyocytes via enhancement of YES-associated protein (YAP)-transcriptional enhancer factor domain (TEAD) activity and improves cardiac function after myocardial infarction in adult mice. To test whether TT-10 can also promote the proliferative capacity of human cardiomyocytes, we investigated the efficacy of TT-10 on human induced pluripotent stem cell (hiPSC)-derived cardiomyocytes (hiPSCMs). The hiPSCs were established from monocytes obtained from healthy donors and cardiac differentiation was performed using a chemically defined protocol. As was observed in murine cardiomyocytes, TT-10 markedly promoted cell cycle activation and increased cell division of hiPSCMs. We then evaluated other effects of TT-10 on the functional properties of hiPSCMs by gene expression and cell motion analyses. We observed that TT-10 had no unfavorable effects on the expression of functional and structural genes or the contractile properties of hiPSCMs. Our results suggest that the novel drug TT-10 effectively activated the cell cycle of hiPSCMs without apparent functional impairment of myocardium, suggesting the potential of clinical usefulness of this drug.