Merck
  • Fabrication of polyphenol-incorporated anti-inflammatory hydrogel via high-affinity enzymatic crosslinking for wet tissue adhesion.
Fabrication of polyphenol-incorporated anti-inflammatory hydrogel via high-affinity enzymatic crosslinking for wet tissue adhesion.
Biomaterials (2020-03-08)
Su-Hwan Kim, Kyungmin Kim, Beom Seok Kim, Young-Hyeon An, Uk-Jae Lee, Sang-Hyuk Lee, Seunghyun L Kim, Byung-Gee Kim, Nathaniel S Hwang
ABSTRACT
Epigallocatechin gallates (EGCGs), isolated from green tea, have intrinsic properties such as anti-oxidant, anti-inflammation, and radical scavenger effects. In this study, we report a tissue adhesive and anti-inflammatory hydrogel formed by high-affinity enzymatic crosslinking of polyphenolic EGCGs. A mixture of EGCG conjugated hyaluronic acids (HA_E) and tyramine conjugated hyaluronic acids (HA_T) was reacted with tyrosinase isolated from Streptomyces avermitillis (SA_Ty) to form that displayed fast enzyme kinetic to form a crosslinked adhesive hydrogel. A 1,2,3-trihydroxyphenyl group in EGCG displayed a high affinity to SA_Ty that allowed HA_E to be quickly oxidized and crosslinked with HA_T to form HA_T and HA_E mixed hydrogel (HA_TE). We then compared the HA_TE hydrogel with commercially available tissue adhesives, such as cyanoacrylate and fibrin glue. We report that the HA_TE exhibited the highest tissue adhesiveness both in wet and dry conditions. Furthermore, HA_TE successfully closed a skin wound and displayed insignificant host tissue responses. This demonstrates that polyphenol-incorporated anti-inflammatory hydrogel may provide a robust tissue adhesive platform for clinical applications.
Sigma-Aldrich
Aminoacetaldehyde diethyl acetal, 98%