Merck
Pharmacokinetics of procainamide and N-acetylprocainamide in rats.
Journal of pharmaceutical sciences (1981-03-01)
B L Kamath, C M Lai, S D Gupta, M J Durrani, A Yacobi
ABSTRACT
The pharmacokinetics of distribution and elimination of procainamide and its major metabolite, N-actylprocainamide, were studied in rats. Eight rats were selected randomly, and each received intravenously 14C-labeled procainamide hydrochloride (75 mg/kg) or 14C-labeled N-acetylprocainamide hydrochloride (86 mg/kg) according to a two-way crossover design. Serial blood samples were withdrawn for 8 hr, and cumulative urine and feces were collected for 48 hr. The plasma concentration-time relationships of procainamide and N-acetylprocainamide were characterized by one- and two-compartment open models, respectively. A pseudo-three-compartment model was necessary to characterize the time course of N-acetylprocainamide in plasma formed after administration of procainamide. The biological half-lives of procainamide and N-acetylprocainamide averaged 0.66 and 2.1 hr, respectively. The urinary excretion profiles of these drugs and the ratio of their biological half-lives in rats were similar to those in humans.
Sigma-Aldrich
N-Acetylprocainamide hydrochloride, ≥99% (HPLC), powder
Pricing and availability is not currently available.