Merck
  • Circulating plasma microRNAs dysregulation and metabolic endotoxemia induced by a high-fat high-saturated diet.
Circulating plasma microRNAs dysregulation and metabolic endotoxemia induced by a high-fat high-saturated diet.
Clinical nutrition (Edinburgh, Scotland) (2019-03-20)
Bruna Jardim Quintanilha, Ludmila Rodrigues Pinto Ferreira, Frederico Moraes Ferreira, Edécio Cunha Neto, Geni Rodrigues Sampaio, Marcelo Macedo Rogero
ABSTRACT
High-fat diet increase two to three times the plasma lipopolysaccharide (LPS) levels and induce subclinical inflammation. Diet can modify gene expression due to epigenetic processes related to MicroRNAs (miRNAs). MicroRNAs (miRNAs) play important role in the post-transcriptional mechanisms involved in regulation of expression of genes related to the inflammatory response. Also, diet can indirectly induce post-transcriptional regulation of gene expression by miRNAs, which may affect the risk for the development of chronic diseases. This study investigated the effect of high-fat high-saturated meal ingestion on plasma miRNA expression and LPS levels during the postprandial period in healthy women. An interventional study was carried out in which a high-fat breakfast (1067.45 kcal), composed mainly of saturated fatty acids (56 g), and 500 mL of water, was offered. Blood samples were collected at baseline and 1, 3 and 5 h after meal intake. The studied population consisted of healthy women (n = 11), aged between 20 and 40 years, and body mass index (BMI) between 18.5 and 25 kg/m2. Plasma levels of lipid profile, cytokines, adhesion molecules, and LPS were measured at the 3 time points. A profile of 752 human plasma miRNA expression was analyzed by real-time PCR assay. These analyzes were performed for all blood collection time-points. Expression profile analysis revealed 33 differentially expressed plasma circulating miRNAs compared to that of the control group. MiR-145-5p and miR-200 were differentially modulated in all time-points post meal consumption. In addition, there was a significant increase in plasma LPS, triglycerides, myristic and palmitic saturated fatty acids levels at the 3 time-points in comparison with the control basal levels. We also observed increased levels of the plasma tumor necrosis factor alpha (TNF-α) cytokine and the vascular cell adhesion molecule 1 (VCAM-1) levels after 5 h post meal ingestion. Ingestion of high-fat high-saturated meal was able to induce metabolic endotoxemia and increase the expression of pro-inflammatory molecules such as TNF-alpha and VCAM-1, as well as modulating circulating miRNAs possibly controlling inflammatory and lipid metabolism proteins at the postprandial period.
Product Description
Supelco
F.A.M.E. Column Evaluation Mix, certified reference material, 1000 μg/mL each component in dichloromethane