Merck
  • Amyloid beta-protein1-42 increases cAMP and apolipoprotein E levels which are inhibited by beta1 and beta2-adrenergic receptor antagonists in mouse primary astrocytes.
Amyloid beta-protein1-42 increases cAMP and apolipoprotein E levels which are inhibited by beta1 and beta2-adrenergic receptor antagonists in mouse primary astrocytes.
Neuroscience (2006-08-15)
U Igbavboa, L N Johnson-Anuna, X Rossello, T A Butterick, G Y Sun, W G Wood
ABSTRACT
Amyloid beta-protein (Abeta) increases apolipoprotein E (apoE) levels in astrocytes which could alter lipid trafficking. The mechanism for the Abeta-induced increase in apoE levels is not well understood. It is well established that stimulation of beta-adrenergic receptors (betaARs) increases cAMP levels. Elevation of cAMP levels increases apoE abundance. The current study determined if Abeta(1-42) stimulation of cAMP and apoE levels could be inhibited by betaAR antagonists in astrocytes. We demonstrate that Abeta(1-42) but not the reverse protein Abeta(42-1) or Abeta(1-40) stimulated cAMP formation and this stimulation was inhibited by selective betaAR antagonists in mouse primary cortical astrocytes. Abeta(1-42) significantly increased apoE levels which were significantly inhibited by the betaAR selective antagonists with the greatest inhibition observed with the beta(2) antagonist. Separate lines of evidence have suggested that agonist-induced stimulation of betaARs and increases in apoE abundance may serve a neuroprotective role in astrocytes. Our results indicate a potential interaction between betaARs and apoE which may contribute to reducing Abeta(1-42) neurotoxicity.
Sigma-Aldrich
Amyloid β 42-1 reverse human, ≥95% (HPLC)