Merck
  • Angiotensin-converting enzyme inhibitors. Mercaptan, carboxyalkyl dipeptide, and phosphinic acid inhibitors incorporating 4-substituted prolines.
Angiotensin-converting enzyme inhibitors. Mercaptan, carboxyalkyl dipeptide, and phosphinic acid inhibitors incorporating 4-substituted prolines.
Journal of medicinal chemistry (1988-06-01)
J Krapcho, C Turk, D W Cushman, J R Powell, J M DeForrest, E R Spitzmiller, D S Karanewsky, M Duggan, G Rovnyak, J Schwartz
ABSTRACT
Analogues of captopril, enalaprilat, and the phosphinic acid [hydroxy(4-phenylbutyl)phosphinyl]acetyl]-L-proline incorporating 4-substituted proline derivatives have been synthesized and evaluated as inhibitors of angiotensin-converting enzyme (ACE) in vitro and in vivo. The 4-substituted prolines, incorporating alkyl, aryl, alkoxy, aryloxy, alkylthio, and arylthio substituents were prepared from derivatives of 4-hydroxy- and 4-ketoproline. In general, analogues of all three classes of inhibitors with hydrophobic substituents on proline were more potent in vitro than the corresponding unsubstituted proline compounds. 4-Substituted analogues of captopril showed greater potency and duration of action than the parent compound as inhibitors of the angiotensin I induced pressor response in normotensive rats. The S-benzoyl derivative of cis-4-(phenylthio)captopril, zofenopril, was found to be one of the most potent compounds of this class and is now being evaluated clinically as an antihypertensive agent. In the phosphinic acid series, the 4-ethylenethioketal and trans-4-cyclohexyl derivatives were found to be the most potent compounds in vitro and in vivo. A prodrug of the latter compound, fosinopril, is also being evaluated in clinical trials.
Sigma-Aldrich
N-Acetyl-L-proline
Pricing and availability is not currently available.