Merck
  • Long-term effects of low doses of Chlorpyrifos exposure at the preweaning developmental stage: A locomotor, pharmacological, brain gene expression and gut microbiome analysis.
Long-term effects of low doses of Chlorpyrifos exposure at the preweaning developmental stage: A locomotor, pharmacological, brain gene expression and gut microbiome analysis.
Food and chemical toxicology : an international journal published for the British Industrial Biological Research Association (2019-10-17)
Cristian Perez-Fernandez, Miguel Morales-Navas, Laia Guardia-Escote, José Antonio Garrido-Cárdenas, María Teresa Colomina, Estela Giménez, Fernando Sánchez-Santed
ABSTRACT
Development is especially sensitive to Chlorpyrifos (CPF) toxicity, associated with several neurodegenerative and neurodevelopmental disorders where motor function dysfunction is a core symptom. Amongst the alternative molecular targets to cholinesterases inhibition, developmental CPF alters different components in the most important neurotransmitter systems, although this depends on the exposure period. Exposure during the late postnatal preweaning stage is the least studied by far. This period includes essential neurodevelopmental processes and has an important translational meaning. The present study analyzed the influence of low doses of CPF on this developmental window on locomotor activity and the state of the different neurotransmitter systems by pharmacological challenges. Brain gene expression and microbiome modulation following CPF were also analyzed. CPF exposure long-term increased spontaneous vertical activity, female's activity following acute stress, hyposensitized the cholinergic system and hypersensitized the GABAergic system, up-regulated both muscarinic 2 receptor and GABA-A-α2 receptor subunit in the dorsal striatum and the frontal cortex, respectively and induced gut microbiota dysbiosis at both genus and species levels. The present study supports alternative molecular targets than the ChEs following late postnatal, preweaning exposure to low doses of CPF, focusing on both cholinergic and GABAergic systems and the gut microbiome as an important factor.
Product Description
Supelco
DL-Amphetamine hydrochloride, analytical standard