Merck
Water-Alcohol Bigels from Fatty Acylated Dipeptides.
The journal of physical chemistry. B (2019-12-28)
Debika Datta, Ramakrishnan Nagaraj, Nitin Chaudhary
ABSTRACT
Peptide-based gels are emerging as an interesting class of biocompatible soft materials. 9-Fluorenylmethoxycarbonyl-protected amino acids and short peptides have gained considerable attention as promising gelators. Peptide amphiphiles, wherein an alkyl chain is appended to a polar peptidic moiety, are another important class of peptide-based gelators. Here, we report the alcohol/water bigels formed by the rather simple fatty acylated dipeptides wherein the peptidic moiety is made up of hydrophobic amino acids, viz., Val, Ile, and Leu. Lauroyl, myristoyl, and palmitoyl were investigated as the N-terminal fatty acyl groups. None of the lauroylated peptides caused gelation of methanol/water and ethanol/water mixtures up to 2 wt % peptide concentration. Eight out of the 27 peptides resulted in distinct bigels. The gels are composed of fibrous aggregates as characterized by electron microscopy. Infrared spectroscopy suggests the β-sheet conformation of the peptidic region in the gels. Using the Ma-IV ethanol/water bigel as the representative gel, entrapment and steady release of the anticancer drug docetaxel are demonstrated. Such bigels from rather simple amphipathic peptides that are easily synthesized and purified through solvent extraction could be attractive gelator candidates with potential application in drug delivery.