Merck
  • Real-time methylation-specific PCR for the evaluation of methylation status of MGMT gene in glioblastoma.
Real-time methylation-specific PCR for the evaluation of methylation status of MGMT gene in glioblastoma.
Oncotarget (2018-07-03)
Masaki Yoshioka, Tomoo Matsutani, Ayaka Hara, Seiichiro Hirono, Takaki Hiwasa, Masaki Takiguchi, Yasuo Iwadate
ABSTRACT
The methylation status of the O6-methylguanine-DNA methyltransferase (MGMT) gene is a strong predictor for the efficacy of temozolomide chemotherapy and survival periods. However, the correlation between the extent of methylation and the difference in survival times has not been fully clarified. Simple and quantitative evaluations of the methylation status in the promotor region of the MGMT gene are expected to be worldwide standardized diagnostics. We applied real-time semi-quantitative methylation-specific polymerase chain reaction (SQ-MSP) of the MGMT gene promoter region to 84 glioblastoma patients. The SQ-MSP result showed that the ΔCt value, which represents the difference between uCt and mCt (uCt value - mCt value), is inversely correlated with overall survival. With adequate cutoff setting, this assay showed that those patients suffering from a tumor with low ΔCt (methylated) survived significantly longer than those having tumors with high ΔCt (un-methylated). The most significant difference was observed when the cutoff was set at a ΔCt of 2. Using this cutoff point, the result of MGMT immunohistochemical analysis was also significantly correlated with the methylation status examined with real-time SQ-MSP. These results collectively show that MGMT promoter methylation status actually affects patients' survival and protein expression depending on its methylation level, and the extent of methylated CpGs would be better assessed with real-time SQ-MSP than with the standard gel-based MSP. This method is cost- and labor-saving compared with pyrosequencing, and significantly contributes to the accurate and objective prediction of patient survival.
Sigma-Aldrich
Anti-MGMT Antibody, clone MT3.1, clone MT3.1, Chemicon®, from mouse
Pricing and availability is not currently available.