Merck
  • The mycotoxin aflatoxin B1 stimulates Epstein-Barr virus-induced B-cell transformation in in vitro and in vivo experimental models.
The mycotoxin aflatoxin B1 stimulates Epstein-Barr virus-induced B-cell transformation in in vitro and in vivo experimental models.
Carcinogenesis (2015-10-02)
Rosita Accardi, Henri Gruffat, Cécilia Sirand, Floriane Fusil, Tarik Gheit, Hector Hernandez-Vargas, Florence Le Calvez-Kelm, Alexandra Traverse-Glehen, François-Loïc Cosset, Evelyne Manet, Christopher P Wild, Massimo Tommasino
ABSTRACT
Although Epstein-Barr virus (EBV) infection is widely distributed, certain EBV-driven malignancies are geographically restricted. EBV-associated Burkitt's lymphoma (eBL) is endemic in children living in sub-Saharan Africa. This population is heavily exposed to food contaminated with the mycotoxin aflatoxin B1 (AFB1). Here, we show that exposure to AFB1 in in vitro and in vivo models induces activation of the EBV lytic cycle and increases EBV load, two events that are associated with an increased risk of eBL in vivo. AFB1 treatment leads to the alteration of cellular gene expression, with consequent activations of signaling pathways, e.g. PI3K, that in turn mediate reactivation of the EBV life cycle. Finally, we show that AFB1 triggers EBV-driven cellular transformation both in primary human B cells and in a humanized animal model. In summary, our data provide evidence for a role of AFB1 as a cofactor in EBV-mediated carcinogenesis.