Merck
  • Copper levels affect targeting of hypoxia-inducible factor 1α to the promoters of hypoxia-regulated genes.
Copper levels affect targeting of hypoxia-inducible factor 1α to the promoters of hypoxia-regulated genes.
The Journal of biological chemistry (2018-08-08)
Xiaojuan Liu, Wenjing Zhang, Zhijuan Wu, Yutao Yang, Y James Kang
ABSTRACT
Hypoxia-inducible factor 1α (HIF-1α) is a transcription factor that regulates cellular responses to hypoxia. It controls the expression of both BCL2/adenovirus E1B 19-kDa protein-interacting protein 3 (BNIP3) and insulin-like growth factor 2 (IGF2). Previous studies have demonstrated that in hypoxia, copper is required for the expression of BNIP3 but not for that of IGF2 Here, using ChIP assays, computational analyses, luciferase reporter assays, and real-time quantitative RT-PCR, we sought to better understand how copper regulates the differential target gene selectivity of HIF-1α. Human umbilical vein endothelial cells (HUVECs) were exposed to CoCl2 or hypoxia conditions to increase HIF-1α accumulation. The binding of HIF-1α to hypoxia-responsive element (HRE) sites in the BNIP3 or IGF2 gene promoter in high- or low-copper conditions was examined. Our analyses revealed three and two potential HRE sites in the BNIP3 and IGF2 promoters, respectively. We identified that HRE (-412/-404) in the BNIP3 promoter and HRE (-354/-347) in the IGF2 promoter are the critical binding sites of HIF-1α. Tetraethelenepentamine (TEPA)-mediated reduction in copper concentration did not affect hypoxia- or CoCl2-induced HIF-1α accumulation. However, the copper reduction did suppress the binding of HIF-1α to the HRE (-412/-404) in BNIP3 but not the binding of HIF-1α to the HRE (-354/-347) in IGF2 In summary, our findings uncovered the mechanistic basis for differential HIF-1α-mediated regulation of BNIP3 and IGF2, indicating that copper regulates target gene selectivity of HIF-1α at least in part by affecting HIF-1α binding to its cognate HRE in the promoters of these two genes.