Merck
  • Histone deacetylase 7 promotes Toll-like receptor 4-dependent proinflammatory gene expression in macrophages.
Histone deacetylase 7 promotes Toll-like receptor 4-dependent proinflammatory gene expression in macrophages.
The Journal of biological chemistry (2013-07-16)
Melanie R Shakespear, Daniel M Hohenhaus, Greg M Kelly, Nabilah A Kamal, Praveer Gupta, Larisa I Labzin, Kate Schroder, Valerie Garceau, Sheila Barbero, Abishek Iyer, David A Hume, Robert C Reid, Katharine M Irvine, David P Fairlie, Matthew J Sweet
ABSTRACT
Broad-spectrum inhibitors of histone deacetylases (HDACs) constrain Toll-like receptor (TLR)-inducible production of key proinflammatory mediators. Here we investigated HDAC-dependent inflammatory responses in mouse macrophages. Of the classical Hdacs, Hdac7 was expressed at elevated levels in inflammatory macrophages (thioglycollate-elicited peritoneal macrophages) as compared with bone marrow-derived macrophages and the RAW264 cell line. Overexpression of a specific, alternatively spliced isoform of Hdac7 lacking the N-terminal 22 amino acids (Hdac7-u), but not the Refseq Hdac7 (Hdac7-s), promoted LPS-inducible expression of Hdac-dependent genes (Edn1, Il-12p40, and Il-6) in RAW264 cells. A novel class IIa-selective HDAC inhibitor reduced recombinant human HDAC7 enzyme activity as well as TLR-induced production of inflammatory mediators in thioglycollate-elicited peritoneal macrophages. Both LPS and Hdac7-u up-regulated the activity of the Edn1 promoter in an HDAC-dependent fashion in RAW264 cells. A hypoxia-inducible factor (HIF) 1 binding site in this promoter was required for HDAC-dependent TLR-inducible promoter activity and for Hdac7- and HIF-1α-mediated trans-activation. Coimmunoprecipitation assays showed that both Hdac7-u and Hdac7-s interacted with HIF-1α, whereas only Hdac7-s interacted with the transcriptional repressor CtBP1. Thus, Hdac7-u positively regulates HIF-1α-dependent TLR signaling in macrophages, whereas an interaction with CtBP1 likely prevents Hdac7-s from exerting this effect. Hdac7 may represent a potential inflammatory disease target.
Sigma-Aldrich
Lipopolysaccharides from Salmonella enterica serotype minnesota, purified by gel-filtration chromatography
Pricing and availability is not currently available.