Merck
  • Efficient syntheses of korupensamines A, B and michellamine B by asymmetric Suzuki-Miyaura coupling reactions.
Efficient syntheses of korupensamines A, B and michellamine B by asymmetric Suzuki-Miyaura coupling reactions.
Journal of the American Chemical Society (2013-10-24)
Guangqing Xu, Wenzhen Fu, Guodu Liu, Chris H Senanayake, Wenjun Tang
ABSTRACT
Efficient asymmetric Suzuki-Miyaura coupling reactions are employed for the first time in total syntheses of chiral biaryl natural products korupensamine A and B in combination with an effective diastereoselective hydrogenation, allowing ultimately a concise and stereoselective synthesis of michellamine B. Chiral monophosphorus ligands L1-3 are effective for the syntheses of a series of functionalized chiral biaryls by asymmetric Suzuki-Miyaura coupling reactions in excellent yields and enantioselectivities (up to 99% ee). The presence of a polar-π interaction between the highly polarized BOP group and the extended π system of arylboronic acid coupling partner is believed to be important for the high enantioselectivity.