Merck
  • Lowered PON1 activities are strongly associated with depression and bipolar disorder, recurrence of (hypo)mania and depression, increased disability and lowered quality of life.
Lowered PON1 activities are strongly associated with depression and bipolar disorder, recurrence of (hypo)mania and depression, increased disability and lowered quality of life.
The world journal of biological psychiatry : the official journal of the World Federation of Societies of Biological Psychiatry (2017-04-27)
Estefania Gastaldello Moreira, Dalmo Guilherme Correia, Kamila Landucci Bonifácio, Juliana Brum de Moraes, Fernanda Liboni Cavicchioli, Carolina Sampaio Nunes, Sandra Odebrecht Vargas Nunes, Heber Odebrecht Vargas, Décio Sabbatini Barbosa, Michael Maes
ABSTRACT
Objectives: Mood disorders (MDs) frequently co-exist with cardiovascular disease (CVD) and immune-inflammatory and oxidative stress are important shared pathophysiological pathways. Even though there has been an extensive investigation of the enzyme paraoxonase 1 (PON1) as a biomarker of susceptibility for CVD, there are few reports studying PON1 in MDs. The aim of this study was to determine the association between PON1 activities as well as functional genotypes and MD diagnosis, clinical characteristics and outcomes. Methods: PON1 activities and functional genotypes were assayed in 58 bipolar disorder (BD) and 32 major depressed patients (MDD) and compared with 59 controls. Results: Our findings show significantly lower PON1 total and CMPAase activities in MDs, which are partly related to the number of previous depressive and manic episodes. Lowered CMPAase activity is associated with a worse outcome of MDs as indicated by lowered quality of life (WHOQoL-BREF scale) and increased disability in the Sheeham scale. Conclusions: We hypothesise that lowered PON1 total and CMPAase activities may play a role in the pathophysiology of MDs by lowering antioxidant defences thereby increasing the risk of lipid peroxidation and inflammation; lowered inhibition of quorum-sensing lactones thereby increasing bacterial proliferation; and attenuated homocysteine thiolactone catabolism which may trigger immune-inflammatory response and/or induce neurotoxicity.