Merck
Emerging biomarkers in ovarian granulosa cell tumors.
International journal of gynecological cancer : official journal of the International Gynecological Cancer Society (2019-03-06)
Anne M Mills, Zachary Chinn, Lisa A Rauh, Anna C Dusenbery, Rachel M Whitehair, Erin Saks, Linda R Duska
ABSTRACT
Although the majority of ovarian granulosa cell tumors can be successfully managed with surgery, a subset require chemotherapy for residual and recurrent disease. The benefit of chemotherapy in this population, however, remains controversial. There is therefore interest in the development of more tolerable and effective treatment options for advanced ovarian granulosa cell tumors. We report the use of immunohistochemistry to investigate how biomarkers could inform clinical trials in granulosa cell tumors with an emphasis on emerging androgen antagonistic, immunotherapeutic, and anti-angiogenic approaches. Immunohistochemistry for androgen receptor, the immune markers programmed cell death ligand 1, indoleamine-2,3 dioxygenase, and cluster of differentiation 8, and the vascular marker cluster of differentiation 31 were evaluated on formalin-fixed paraffin-embedded whole tissue sections from 29 cases of adult-type granulosa cell tumors. Results were evaluated with clinicopathologic variables including recurrence. 59% of granulosa cell tumors were androgen receptor-positive, suggesting a potential role for anti-androgen therapy in this tumor type. In contrast, the targetable immune modulatory molecules programmed cell death ligand 1 and indoleamine-2,3 dioxygenase were scarcely expressed, with no cases showing tumorous programmed cell death ligand 1 and a single case demonstrating very focal tumorous indoleamine-2,3 dioxygenase staining. A minority of cases expressed programmed cell death ligand 1 in occasional tumor-associated macrophages and indoleamine-2,3 dioxygenase in peritumoral vessels. Tumor-infiltrating cytotoxic T cells were also scarce in granulosa cell tumors, arguing against a significant role for immunotherapy in the absence of additional immunostimulation. Cluster of differentiation 31 immunostaining revealed a range of vascular densities across granulosa cell tumors, and future studies evaluating the role of vascular density as a predictor of response to angiogenesis inhibition are warranted. None of the biomarkers investigated were significantly correlated with recurrence, and the only clinicopathologic feature significantly correlated with outcome was stage at presentation. Biomarker data suggest that many ovarian granulosa cell tumors could be candidates for anti-androgen therapy, while the potential role for immunotherapy appears more limited. Vascular density could be useful for identifying optimal candidates for angiogenesis inhibition. Incorporation of these biomarkers into clinical trials could help optimize patient selection.