Merck
  • Nitric oxide activation of poly(ADP-ribose) synthetase in neurotoxicity.
Nitric oxide activation of poly(ADP-ribose) synthetase in neurotoxicity.
Science (New York, N.Y.) (1994-02-04)
J Zhang, V L Dawson, T M Dawson, S H Snyder
ABSTRACT
Poly(adenosine 5'-diphosphoribose) synthetase (PARS) is a nuclear enzyme which, when activated by DNA strand breaks, adds up to 100 adenosine 5'-diphosphoribose (ADP-ribose) units to nuclear proteins such as histones and PARS itself. This activation can lead to cell death through depletion of beta-nicotinamide adenine dinucleotide (the source of ADP-ribose) and adenosine triphosphate. Nitric oxide (NO) stimulated ADP-ribosylation of PARS in rat brain. Benzamide and other derivatives, which inhibit PARS, blocked N-methyl-D-aspartate- and NO-mediated neurotoxicity with relative potencies paralleling their ability to inhibit PARS. Thus, NO appeared to elicit neurotoxicity by activating PARS.
Sigma-Aldrich
1,5-Isoquinolinediol, ≥98% (HPLC), powder