Merck
  • Myoglobinopathy is an adult-onset autosomal dominant myopathy with characteristic sarcoplasmic inclusions.
Myoglobinopathy is an adult-onset autosomal dominant myopathy with characteristic sarcoplasmic inclusions.
Nature communications (2019-03-29)
Montse Olivé, Martin Engvall, Gianina Ravenscroft, Macarena Cabrera-Serrano, Hong Jiao, Carlo Augusto Bortolotti, Marcello Pignataro, Matteo Lambrughi, Haibo Jiang, Alistair R R Forrest, Núria Benseny-Cases, Stefan Hofbauer, Christian Obinger, Gianantonio Battistuzzi, Marzia Bellei, Marco Borsari, Giulia Di Rocco, Helena M Viola, Livia C Hool, Josep Cladera, Kristina Lagerstedt-Robinson, Fengqing Xiang, Anna Wredenberg, Francesc Miralles, Juan José Baiges, Edoardo Malfatti, Norma B Romero, Nathalie Streichenberger, Christophe Vial, Kristl G Claeys, Chiara S M Straathof, An Goris, Christoph Freyer, Martin Lammens, Guillaume Bassez, Juha Kere, Paula Clemente, Thomas Sejersen, Bjarne Udd, Noemí Vidal, Isidre Ferrer, Lars Edström, Anna Wedell, Nigel G Laing
ABSTRACT
Myoglobin, encoded by MB, is a small cytoplasmic globular hemoprotein highly expressed in cardiac myocytes and oxidative skeletal myofibers. Myoglobin binds O2, facilitates its intracellular transport and serves as a controller of nitric oxide and reactive oxygen species. Here, we identify a recurrent c.292C>T (p.His98Tyr) substitution in MB in fourteen members of six European families suffering from an autosomal dominant progressive myopathy with highly characteristic sarcoplasmic inclusions in skeletal and cardiac muscle. Myoglobinopathy manifests in adulthood with proximal and axial weakness that progresses to involve distal muscles and causes respiratory and cardiac failure. Biochemical characterization reveals that the mutant myoglobin has altered O2 binding, exhibits a faster heme dissociation rate and has a lower reduction potential compared to wild-type myoglobin. Preliminary studies show that mutant myoglobin may result in elevated superoxide levels at the cellular level. These data define a recognizable muscle disease associated with MB mutation.