Merck
  • Type 3 inositol 1,4,5-trisphosphate receptor has antiapoptotic and proliferative role in cancer cells.
Type 3 inositol 1,4,5-trisphosphate receptor has antiapoptotic and proliferative role in cancer cells.
Cell death & disease (2019-02-24)
Ingeborg Rezuchova, Sona Hudecova, Andrea Soltysova, Miroslava Matuskova, Erika Durinikova, Barbora Chovancova, Michal Zuzcak, Marina Cihova, Monika Burikova, Adela Penesova, Lubomira Lencesova, Jan Breza, Olga Krizanova
ABSTRACT
Although the involvement of type 1 (IP3R1) and type 2 (IP3R2) inositol 1,4,5-trisphosphate receptors in apoptosis induction has been well documented in different cancer cells and tissues, the function of type 3 IP3R (IP3R3) is still elusive. Therefore, in this work we focused on the role of IP3R3 in tumor cells in vitro and in vivo. We determined increased expression of this receptor in clear cell renal cell carcinoma compared to matched unaffected part of the kidney from the same patient. Thus, we hypothesized about different functions of IP3R3 compared to IP3R1 and IP3R2 in tumor cells. Silencing of IP3R1 prevented apoptosis induction in colorectal cancer DLD1 cells, ovarian cancer A2780 cells, and clear cell renal cell carcinoma RCC4 cells, compared to apoptosis in cells treated with scrambled siRNA. As expected, silencing of IP3R3 and subsequent apoptosis induction resulted in increased levels of apoptosis in all these cells. Further, we prepared a DLD1/IP3R3_del cell line using CRISPR/Cas9 gene editing method. These cells were injected into nude mice and tumor's volume was compared with tumors induced by DLD1 cells. Lower volume of tumors originated from DLD1/IP3R3_del cells was observed after 12 days, compared to wild type DLD1 cells. Also, the migration of these cells was lesser compared to wild type DLD1 cells. Apoptosis under hypoxic conditions was more pronounced in DLD1/IP3R3_del cells than in DLD1 cells. These results clearly show that IP3R3 has proliferative and anti-apoptotic effect in tumor cells, on contrary to the pro-apoptotic effect of IP3R1.
Sigma-Aldrich
Anti-IP₃ Receptor Mouse mAb (IPR.1), liquid, clone IPR.1, Calbiochem®