Merck
  • L-homocysteic acid: an endogenous excitotoxic ligand of the NMDA receptor.
L-homocysteic acid: an endogenous excitotoxic ligand of the NMDA receptor.
Brain research bulletin (1987-11-01)
J W Olney, M T Price, K S Salles, J Labruyere, R Ryerson, K Mahan, G Frierdich, L Samson
ABSTRACT
L-Homocysteic acid (L-HCA) has been proposed as a natural transmitter at the N-methyl-D-aspartate (NMDA) subtype of excitatory amino acid receptor based on recent evidence that L-HCA occurs L-HCA occurs naturally in the mammalian CNS, is released from K+ stimulated brain slices in a calcium-dependent manner and may be contained in nerve terminals located in certain brain regions that have a high density of NMDA receptors. Here we report that L-HCA potently induces a pattern of cytopathology in the ex vivo chick retina which mimics the pattern of NMDA but not kainic acid (KA) neurotoxicity. We also show that known NMDA antagonists, including Mg++, D-aminophosphonopentanoate and certain anesthetics, analgesics, and sedative hypnotics block the neurotoxic actions of L-HCA in direct proportion to their efficacy in blocking NMDA neurotoxicity. While there is a perfect correspondence between agents that block NMDA and L-HCA neurotoxicity, only a few such agents are active against KA neurotoxicity. We find that 3H-Glu binding is inhibited more potently by L-HCA (Ki = 67 microM). Moreover the patterns with which L-HCA and NMDA displace 3H-Glu binding in autoradiograms appear essentially identical. These findings are consistent with the proposal that L-HCA is an endogenous ligand at NMDA receptors.
Sigma-Aldrich
L-Homocysteic acid, ≥95%
Pricing and availability is not currently available.