- Protein structure determination using a combination of cross-linking, mass spectrometry, and molecular modeling.
Protein structure determination using a combination of cross-linking, mass spectrometry, and molecular modeling.
Methods in molecular biology (Clifton, N.J.) (2008-06-11)
Dmitri Mouradov, Gordon King, Ian L Ross, Jade K Forwood, David A Hume, Andrea Sinz, Jennifer L Martin, Bostjan Kobe, Thomas Huber
PMID18542884
ABSTRACT
Cross-linking in combination with mass spectrometry can be used as a tool for structural modeling of protein complexes and multidomain proteins. Although cross-links represent only weak structural constraints, the combination of a limited set of experimental cross-links with molecular docking/modeling is often sufficient to determine the structure of a protein complex or multidomain protein at low resolution.
Product Description
Supelco
ProteoMass™ Peptide and Protein MALDI-MS Calibration Kit, Peptide and Protein MALDI MS Calibration Standard