Merck
  • Isodesmic C-H Borylation: Perspectives and Proof of Concept of Transfer Borylation Catalysis.
Isodesmic C-H Borylation: Perspectives and Proof of Concept of Transfer Borylation Catalysis.
Journal of the American Chemical Society (2019-07-10)
Étienne Rochette, Vincent Desrosiers, Yashar Soltani, Frédéric-Georges Fontaine
ABSTRACT
The potential advantages of using arylboronic esters as boron sources in C-H borylation are discussed. The concept is showcased using commercially available 2-mercaptopyridine as a metal-free catalyst for the transfer borylation of heteroarenes using arylboronates as borylation agents. The catalysis shows a unique functional group tolerance among C-H borylation reactions, tolerating notably terminal alkene and alkyne functional groups. The mechanistic investigation is also described.
Sigma-Aldrich
2-Mercaptopyridine, ReagentPlus®, 99%