- Insulin-like growth factor 1 regulates developing brain glucose metabolism.
Insulin-like growth factor 1 regulates developing brain glucose metabolism.
The brain has enormous anabolic needs during early postnatal development. This study presents multiple lines of evidence showing that endogenous brain insulin-like growth factor 1 (Igf1) serves an essential, insulin-like role in promoting neuronal glucose utilization and growth during this period. Brain 2-deoxy-d- [1-(14)C]glucose uptake parallels Igf1 expression in wild-type mice and is profoundly reduced in Igf1-/- mice, particularly in those structures where Igf1 is normally most highly expressed. 2-Deoxy-d- [1-(14)C]glucose is significantly reduced in synaptosomes prepared from Igf1-/- brains, and the deficit is corrected by inclusion of Igf1 in the incubation medium. The serine/threonine kinase Akt/PKB is a major target of insulin-signaling in the regulation of glucose transport via the facilitative glucose transporter (GLUT4) and glycogen synthesis in peripheral tissues. Phosphorylation of Akt and GLUT4 expression are reduced in Igf1-/- neurons. Phosphorylation of glycogen synthase kinase 3beta and glycogen accumulation also are reduced in Igf1-/- neurons. These data support the hypothesis that endogenous brain Igf1 serves an anabolic, insulin-like role in developing brain metabolism.