Merck
  • Mesostructured organosilica with a 9-mesityl-10-methylacridinium bridging unit: photoinduced charge separation in the organosilica framework.
Mesostructured organosilica with a 9-mesityl-10-methylacridinium bridging unit: photoinduced charge separation in the organosilica framework.
Chemical communications (Cambridge, England) (2010-11-03)
Norihiro Mizoshita, Ken-Ichi Yamanaka, Toyoshi Shimada, Takao Tani, Shinji Inagaki
ABSTRACT
Polycondensation of 9-mesityl-10-methylacridinium-bridged organosilane in the presence of a nonionic surfactant yielded a mesostructured organosilica solid with a functional framework that exhibited long-lived photoinduced charge separation.