Merck
  • A New Portal to SuFEx Click Chemistry: A Stable Fluorosulfuryl Imidazolium Salt Emerging as an "F-SO2 +" Donor of Unprecedented Reactivity, Selectivity, and Scope.
A New Portal to SuFEx Click Chemistry: A Stable Fluorosulfuryl Imidazolium Salt Emerging as an "F-SO2 +" Donor of Unprecedented Reactivity, Selectivity, and Scope.
Angewandte Chemie (International ed. in English) (2017-12-26)
Taijie Guo, Genyi Meng, Xiongjie Zhan, Qian Yang, Tiancheng Ma, Long Xu, K Barry Sharpless, Jiajia Dong
ABSTRACT
Sulfuryl fluoride, SO2 F2 , has been found to derivatize phenols in all kinds of environments, even those in highly functional molecules. We now report that a solid fluorosulfuryl imidazolium triflate salt delivers the same "F-SO2 +" fragment to Nu-H acceptor groups in the substrates. However, this triflate salt is a far more reactive fluorosulfurylating agent than SO2 F2 and displays selectivity preferences of its own. Moreover, the new azolium triflate reagent reacts once with primary amines and anilines before the reaction stops. On the other hand, with triethylamine and two equivalents of the "F-SO2 +" donor present, it proceeds on to the bis(fluorosulfuryl)imides in good yield-two important conversions that we have never seen with sulfuryl fluoride as the electrophile.
Sigma-Aldrich
1-(Fluorosulfonyl)-2,3-dimethyl-1H-imidazol-3-ium trifluoromethanesulfonate, ≥95%
Pricing and availability is not currently available.