Merck
  • In vitro propagation of threatened terrestrial orchid, Malaxis khasiana Soland ex. Swartz through immature seed culture.
In vitro propagation of threatened terrestrial orchid, Malaxis khasiana Soland ex. Swartz through immature seed culture.
Indian journal of experimental biology (2006-09-27)
Chitta Ranjan Deb, Temjensangba
ABSTRACT
Rapid in vitro propagation of the terrestrial orchid, M. khasiana through immature seed culture was achieved. Immature seeds of 8-9 week after pollination (WAP) cultured on MS medium (2% sucrose) supplemented with 500 mgl(-1) casein-hydrolysate and 1 microM N6-benzyladenine (BA) exhibited germination of 75% seeds after 107 days of culture and subsequently supported the development of PLBs. Subsequent culture on MS medium enriched with 6 microM of indole-3-acetic acid (IAA), 18 microM each of BA and kinetin induced multiple shoots and plantlets. Transfer of PLBs to MS medium with 0.1% activated charcoal (AC) facilitated rapid proliferation of PLBs, while AC at 0.2% favored shoot bud induction and rhizome enlargement. The plantlets, developed on medium with IAA, BA and kinetin, after hardening in vitro for 8-10 weeks were planted in community pots and transferred to poly-house. The plantlets showed 65% survival under field conditions.
Sigma-Aldrich
3-Indoleacetic acid, suitable for plant cell culture, crystalline