Merck
  • Brain regions and monoaminergic neurotransmitters that are involved in mouse ambulatory activity promoted by bupropion.
Brain regions and monoaminergic neurotransmitters that are involved in mouse ambulatory activity promoted by bupropion.
Toxicology reports (2016-06-21)
Toyoshi Umezu, Yasuyuki Shibata
ABSTRACT
Bupropion (BUP), a substituted phenyl-ethylamine, has been utilized for the treatment of depression and for smoking cessation, however, one concern is that BUP may increase a risk of psychosis similar to other substituted phenyl-ethylamine amphetamine (AMPH) and methamphetamine (MetAMPH). BUP promotes ambulation in mice and causes behavioral sensitization on the ambulation-promoting effect when repeatedly administered as well as AMPH and MetAMPH. The present study aimed to elucidate brain regions and monoaminergic neurotransmitters that are involved in the ambulation-promoting effect of BUP. c-Fos-like immunoreactivity (c-Fos-IR) mapping in brain in combination with measuring ambulatory activity was conducted to determine brain region(s) that is involved in the ambulatory effect of BUP. Three kinds of statistical analyses for c-Fos-IR in 24 brain regions consistently showed that c-Fos-IR in the Caudate putamen (CPu) is positively correlated with the ambulatory response to BUP. In addition, multiple regression analysis indicated that the ambulatory response is a function of c-Fos-IR not only in the CPu but also in the lateral septum nucleus (LS), median raphe nucleus (MnR), lateral globus pallidus (LGP), medial globus pallidus (MGP), locus coeruleus (LC) and ventral hypothalamic nucleus (VMH). Effects of BUP on monoaminergic neurotransmitters in the CPu were examined using in vivo microdialysis method, as the pharmacological experiments indicated that monoaminergic neurotransmitters, dopamine (DA) in particular, mediate the ambulatory response to BUP. Response of DA in the CPu to BUP was parallel to the ambulatory response, showing that DA in the CPu is involved in the ambulatory response to BUP. The present study also suggests that other brain regions such as the LC, the origin nucleus of norepinephrine (NE) neurons, and another neurotransmitter NE may also play some roles for the ambulatory response to BUP, however, further studies are needed to elucidate the roles.
Sigma-Aldrich
Anti-c-Fos antibody produced in rabbit, IgG fraction of antiserum, buffered aqueous solution
Pricing and availability is not currently available.