Merck
Proteolytic control of the mitochondrial calcium uniporter complex.
Proceedings of the National Academy of Sciences of the United States of America (2017-04-12)
Chen-Wei Tsai, Yujiao Wu, Ping-Chieh Pao, Charles B Phillips, Carole Williams, Christopher Miller, Matthew Ranaghan, Ming-Feng Tsai
ABSTRACT
The mitochondrial calcium uniporter is a Ca2+-activated Ca2+ channel complex mediating mitochondrial Ca2+ uptake, a process crucial for Ca2+ signaling, bioenergetics, and cell death. The uniporter is composed of the pore-forming MCU protein, the gatekeeping MICU1 and MICU2 subunits, and EMRE, a single-pass membrane protein that links MCU and MICU1 together. As a bridging subunit required for channel function, EMRE could paradoxically inhibit uniporter complex formation if expressed in excess. Here, we show that mitochondrial mAAA proteases AFG3L2 and SPG7 rapidly degrade unassembled EMRE using the energy of ATP hydrolysis. Once EMRE is incorporated into the complex, its turnover is inhibited >15-fold. Protease-resistant EMRE mutants produce uniporter subcomplexes that induce constitutive Ca2+ leakage into mitochondria, a condition linked to debilitating neuromuscular disorders in humans. The results highlight the dynamic nature of uniporter subunit assembly, which must be tightly regulated to ensure proper mitochondrial responses to intracellular Ca2+ signals.
Sigma-Aldrich
Anti-MCU antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
Sigma-Aldrich
ANTI-FLAG® M2 antibody, Mouse monoclonal, 1 mg/mL, clone M2, affinity isolated antibody, buffered aqueous solution (50% glycerol, 10 mM sodium phosphate, and 150 mM NaCl, pH 7.4)