
Zinc trifluoroethanesulfinate

Synonym(s):
Baran TFES Reagent, Baran Trifluoroethylation Reagent, Bis(((2,2,2-trifluoroethyl)sulfinyl)oxy)zinc, TFES
Empirical Formula (Hill Notation):
C4H4F6O4S2Zn
CAS Number:
Molecular Weight:
359.58
Products
Brand
Formula Weight
Melting Point (°C)
Physical Form
Functional Group
Reaction Type
Reagent Type
Showing 1-1 of 1
Compare
Product Number
Description
Pricing