Skip to Content
Merck
Z-DL-Ser-OH 99%

Z-DL-Ser-OH

Z-DL-Ser-OH 99%
Synonym(s):
N-Cbz-DL-serine
Linear Formula:
HOCH2CH(NHCO2CH2C6H5)CO2H
CAS Number:
Molecular Weight:
239.22
EC Number:

Products

Product Category
Brand
Color
Formula Weight
Melting Point (°C)
Physical Form
Purity
Application
Markush Class
Markush Group
Optical Isomer
Amino Acid
Amino Acid Type
Reaction Type
Showing 1-1 of 1
Compare
Product Number
Description
Pricing