Skip to Content
Merck
Z-DL-Lys(Z)-OH ≥98.0% (TLC)

Z-DL-Lys(Z)-OH

Z-DL-Lys(Z)-OH ≥98.0% (TLC)
Synonym(s):
Nα,Nε-Di-Z-DL-lysine, Nα,Nε-di-Z-DL-lysine
Empirical Formula (Hill Notation):
C22H26N2O6
CAS Number:
Molecular Weight:
414.45

Products

Brand
Formula Weight
Melting Point (°C)
Physical Form
Purity
Application
Markush Class
Markush Group
Optical Isomer
Amino Acid
Amino Acid Type
Reaction Type
Showing 1-1 of 1
Compare
Product Number
Description
Pricing