Skip to Content
Merck
Yttrium(III) tris(2,2,6,6-tetramethyl-3,5-heptanedionate) 99.9% trace metals basis

Yttrium(III) tris(2,2,6,6-tetramethyl-3,5-heptanedionate)

Yttrium(III) tris(2,2,6,6-tetramethyl-3,5-heptanedionate) 99.9% trace metals basis
Synonym(s):
Y(TMHD)3
Linear Formula:
Y(OCC(CH3)3CHCOC(CH3)3)3
CAS Number:
Molecular Weight:
638.71

Products

Brand
Formula Weight
Melting Point (°C)
Physical Form
Purity
Markush Class
Markush Group
Reagent Type
Core
Showing 1-1 of 1