Vanadium(V) oxyfluoride
Synonym(s):
Vanadium(V) oxytrifluoride, Vanadyl trifluoride
Linear Formula:
VOF3
Products
Brand
Formula Weight
Physical Form
Purity
Quality Segment
Application
Reagent Type
Core
Showing 1-1 of 1
Compare
Product Number
Description
Pricing