Skip to Content
Merck
Tris(ethylmethylamido)(tert-butylimido)tantalum(V) electronic grade, ≥95% (CP), ≥99.99% trace metals basis

Tris(ethylmethylamido)(tert-butylimido)tantalum(V)

Tris(ethylmethylamido)(tert-butylimido)tantalum(V) electronic grade, ≥95% (CP), ≥99.99% trace metals basis
Empirical Formula (Hill Notation):
C13H33N4Ta
CAS Number:
Molecular Weight:
426.38

Products

Brand
Formula Weight
Physical Form
Purity
Markush Class
Markush Group
Reagent Type
Core