Skip to Content
Merck
Tris(dimethylamido)antimony(III) 99.99% trace metals basis

Tris(dimethylamido)antimony(III)

Tris(dimethylamido)antimony(III) 99.99% trace metals basis
Synonym(s):
Tris(dimethylamino)antimony(III)
Linear Formula:
[(CH3)2N]3Sb
CAS Number:
Molecular Weight:
253.99

Products

Brand
Boiling Point (°C)
Formula Weight
Physical Form
Purity
Reagent Type
Core
Showing 1-1 of 1
Compare
Product Number
Description
Pricing