Skip to Content
Merck
Tributyl(4,5-dihydrofuran-2-yl)stannane 97%

Tributyl(4,5-dihydrofuran-2-yl)stannane

Tributyl(4,5-dihydrofuran-2-yl)stannane 97%
Synonym(s):
Tributyl(4,5-dihydrofuran-2-yl)tin
Empirical Formula (Hill Notation):
C16H32OSn
CAS Number:
Molecular Weight:
359.13

Products

Brand
Formula Weight
Physical Form
Purity
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing