Skip to Content
Merck
trans-2-Aminocyclohexanecarboxylic acid ≥97.0%

trans-2-Aminocyclohexanecarboxylic acid

trans-2-Aminocyclohexanecarboxylic acid ≥97.0%
Empirical Formula (Hill Notation):
C7H13NO2
CAS Number:
Molecular Weight:
143.18

Products

Product Category
Brand
Color
Formula Weight
Melting Point (°C)
Physical Form
Purity
Quality Segment
Application
Markush Class
Markush Group
Amino Acid Type
Reaction Type
Showing 1-1 of 1
Compare
Product Number
Description
Pricing